Learn More
Biolegend PE/Dazzle™ 594 anti-human CD127 (IL-7Rα)
Supplier:  Biolegend 351336
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD127 (IL-7Rα) [A019D5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-712-126