Learn More
Biolegend PE/Dazzle™ 594 anti-human CD16
Supplier:  Biolegend 302053
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. 50-207-2823