Learn More
Biolegend PE/Dazzle™ 594 anti-human CD169 (Sialoadhesin, Siglec-1)
Supplier:  Biolegend 346016
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD169 (Sialoadhesin, Siglec-1) [7-239]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-6794