Learn More
Biolegend PE/Dazzle™ 594 anti-human CD185 (CXCR5)
Supplier:  Biolegend 356927
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD185 (CXCR5) [J252D4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-712-154