Learn More
Biolegend PE/Dazzle™ 594 anti-human CD196 (CCR6)
Supplier:  Biolegend 353429
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD196 (CCR6) [G034E3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-7252