Learn More
Biolegend PE/Dazzle™ 594 anti-human CD197 (CCR7)
Supplier:  Biolegend 353236
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD197 (CCR7) [G043H7]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-712-131