Learn More
Biolegend PE/Dazzle™ 594 anti-human CD1a
Supplier:  Biolegend 300132
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD1a [HI149]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-2648