Learn More
Biolegend PE/Dazzle™ 594 anti-human CD1c
Supplier:  Biolegend 331531
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-103-0713