Learn More
Biolegend PE/Dazzle™ 594 anti-human CD21
Supplier:  Biolegend 354921
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD21 [Bu32]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-207-7398