Learn More
Biolegend PE/Dazzle™ 594 anti-human CD314 (NKG2D)
Supplier:  Biolegend 320828
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD314 (NKG2D) [1D11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-207-3830