Learn More
Biolegend PE/Dazzle™ 594 anti-human CD45
Supplier:  Biolegend 304052
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-711-989