Learn More
Biolegend PE/Dazzle™ 594 anti-human CD45RA
Supplier:  Biolegend 304146
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-207-3053