Learn More
Biolegend PE/Dazzle™ 594 anti-human IL-2
Supplier:  Biolegend 500343
Encompass_Preferred
PE/Dazzle™ 594 anti-human IL-2 [MQ1-17H12]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cat (Feline), Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Sooty Mangabey; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-5486