Learn More
Biolegend PE/Dazzle™ 594 anti-human Perforin
Supplier:  Biolegend 308131
Encompass_Preferred
PE/Dazzle™ 594 anti-human Perforin [dG9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Bovine (Cow, Cattle); Apps: ICFC; Size: 25 tests
Catalog No. 50-207-3372