Learn More
Biolegend PE/Dazzle™ 594 anti-human TNF-α
Supplier:  Biolegend 502946
Encompass_Preferred
PE/Dazzle™ 594 anti-human TNF-α [MAb11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cat (Feline), Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine), Pigtailed Macaque, Sooty Mangabey; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-850-987