Learn More
Biolegend PE/Dazzle™ 594 anti-human CD110
Supplier:  Biolegend 393705
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD110 [S16017E]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9037