Learn More
Biolegend PE/Dazzle™ 594 anti-human CD19
Supplier:  Biolegend 363031
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8120