Learn More
Biolegend PE/Dazzle™ 594 anti-human CD223 (LAG-3)
Supplier:  Biolegend 369331
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD223 (LAG-3) [11C3C65]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8502