Learn More
Biolegend PE/Dazzle™ 594 anti-human CD279 (PD-1)
Supplier:  Biolegend 367433
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD279 (PD-1) [NAT105]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8354