Learn More
Biolegend PE/Dazzle™ 594 anti-human KLRG1 (MAFA)
Supplier:  Biolegend 368607
Encompass_Preferred
PE/Dazzle™ 594 anti-human KLRG1 (MAFA) [14C2A07]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8446