Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD16.2 (FcγRIV)
Supplier:  Biolegend 149520
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD16.2 (FcγRIV) [9E9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1874