Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD21/CD35 (CR2/CR1)
Supplier:  Biolegend 123439
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD21/CD35 (CR2/CR1) [7E9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0530