Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD11c
Supplier:  Biolegend 117347
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-711-850