Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD134 (OX-40)
Supplier:  Biolegend 119418
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD134 (OX-40) [OX-86]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0354