Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD279 (PD-1)
Supplier:  Biolegend 109115
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD279 (PD-1) [RMP1-30]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0073