Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD43 Activation-Associated Glycoform
Supplier:  Biolegend 121226
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD43 Activation-Associated Glycoform [1B11]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0441