Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD90.2 (Thy1.2) [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Supplier:  Biolegend 105340
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD90.2 (Thy1.2) [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-237-4642