Learn More
Biolegend PE/Dazzle™ 594 anti-mouse/human CD44
Supplier:  Biolegend 103056
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse/human CD44 [IM7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Squirrel Monkey, Horse (Equine), Cattle (Bovine, Cow), Swine (Pig, Porcine), Dog (Canine), Cat (Feline); Apps: FC; Size: 100 ug
Catalog No. 50-850-270