Learn More
Biolegend PE/Dazzle™ 594 anti-mouse IL-10
Supplier:  Biolegend 505034
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-208-5671