Learn More
Biolegend PE/Dazzle™ 594 anti-mouse Ly-6A/E (Sca-1)
Supplier:  Biolegend 108137
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse Ly-6A/E (Sca-1) [D7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0012