Learn More
Biolegend PE/Dazzle™ 594 anti-mouse/rat CD29
Supplier:  Biolegend 102231
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse/rat CD29 [HMβ1-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 25 ug
Catalog No. 50-206-9641