Learn More
Biolegend PE/Dazzle™ 594 Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400557
Encompass_Preferred
PE/Dazzle™ 594 Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 25 ug
Catalog No. 50-712-224