Learn More
Biolegend PE/Dazzle™ 594 Syrian Hamster IgG Isotype Ctrl
Supplier:  Biolegend 402031
Encompass_Preferred
PE/Dazzle™ 594 Syrian Hamster IgG Isotype Ctrl [SHG-1]; Isotype: Syrian Hamster IgG; Apps: FC, ICFC; Size: 25 ug
Catalog No. 50-207-9657