Learn More
Biolegend PE/Fire™ 640 anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 ug
Supplier:  Biolegend 101279
Encompass_Preferred
PE/Fire™ 640 anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 ug
Catalog No. 50-237-8301