Learn More
Biolegend PE/Fire™ 810 anti-human CD194 (CCR4)
Supplier:  Biolegend 359433
Encompass_Preferred
PE/Fire™ 810 anti-human CD194 (CCR4) [L291H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-213-1959