Learn More
Biolegend PE/Fire™ 810 anti-human CD28
Supplier:  Biolegend 302971
Encompass_Preferred
PE/Fire™ 810 anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. 50-213-1863