Learn More
Biolegend PE/Fire™ 810 anti-human CD39
Supplier:  Biolegend 328245
Encompass_Preferred
PE/Fire™ 810 anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-213-1913