Learn More
Biolegend PE/Fire™ 640 anti-mouse CD3
Supplier:  Biolegend 100270
Encompass_Preferred
PE/Fire™ 640 anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-206-9444