Learn More
Biolegend PE/Fire™ 640 anti-mouse CD4
Supplier:  Biolegend 100481
Encompass_Preferred
PE/Fire™ 640 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-206-9480