Learn More
Biolegend PE/Fire™ 700 anti-human CD45
Supplier:  Biolegend 304078
Encompass_Preferred
PE/Fire™ 700 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-207-3050