Learn More
Biolegend PE/Fire™ 700 anti-mouse CD45
Supplier:  Biolegend 103177
Encompass_Preferred
PE/Fire™ 700 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-206-9720