Learn More
Biolegend PE/FireTM 700 anti-mouse NK-
Supplier:  Biolegend 108773
Encompass_Preferred
PE/Fire™ 700 anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-253-8007