Learn More
Biolegend PE/FireTM 744 anti-mouse CD127
Catalog No. 502796415
Encompass_Preferred
PE/Fire™ 744 anti-mouse CD127 (IL-7Rα) [S18006K]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-279-6415 Supplier Biolegend Supplier No. 158217
PE/Fire™ 744 anti-mouse CD127 (IL-7Rα) [S18006K]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg