Learn More
Biolegend PE/FireTM 744 anti-mouse CD279
Supplier:  Biolegend 135269
Encompass_Preferred
PE/Fire™ 744 anti-mouse CD279 (PD-1) [29F.1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-279-6480