Learn More
Biolegend PE/FireTM 744 anti-mouse CD279
Catalog No. 502796557
Encompass_Preferred
PE/Fire™ 744 anti-mouse CD279 (PD-1) [29F.1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-279-6557 Supplier Biolegend Supplier No. 135270
PE/Fire™ 744 anti-mouse CD279 (PD-1) [29F.1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg