Learn More
Biolegend PE/FireTM 744 Streptavidin
Catalog No. 502865934
Encompass_Preferred
PE/Fire™ 744 Streptavidin; Reactivity: Human, Mouse, Rat, All Species; Apps: FC; Size: 100 μg
Catalog No. 50-286-5934 Supplier Biolegend Supplier No. 405388
PE/Fire™ 744 Streptavidin; Reactivity: Human, Mouse, Rat, All Species; Apps: FC; Size: 100 μg