Learn More
Biolegend PE Goat IgG Isotype Ctrl
Supplier:  Biolegend 403004
Encompass_Preferred
PE Goat IgG Isotype Ctrl [Poly24030]; Isotype: Goat Polyclonal IgG; Reactivity: Goat; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-169-073