Learn More
Cytek Biosciences Inc PE HU CD56 (NCAM) (MY31) 100T
Supplier:  Cytek Biosciences Inc 500564T100
Encompass_Preferred
PE Anti-Human CD56 (NCAM) (MY31) Isotype: Mouse IgG1, kappa Apps: FC Size: 100 tests
Catalog No. 50-105-4717