Learn More
ACROBiosystems PE-Labeled Human BCMA / TNFRSF17 Protein, His Tag
Supplier:  ACROBiosystems BCAHP2H225TESTS
Encompass_Preferred
Recombinant Protein;25TESTS;PE-Labeled Human BCMA / TNFRSF17 Protein, His Tag (Cat. No. BCA-HP2H2) is produced via conjugation of PE to Human BCMA / TNFRSF17 Protein, His Tag under optimal conditions. Human BCMA / TNFRSF17 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10 kDa. ;1e6 of the Anti-BCMA CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human BCMA Protein, His Tag (Cat. No. BCA-HP2H2) and negative control protein respectively, PE signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-205-5160