Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human EGF R Protein, His Tag (Site-specific conjugation)
Catalog No. 502106197
Encompass_Preferred
Recombinant Protein;200TESTS ;PE-Labeled Human EGF R, His Tag (EGR-HP2E3) is produced via site-specific conjugation of PE to Human EGF R (25-645), His Tag under optimal conditions with a proprietary technology. Human EGF R (25-645), His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.3 kDa.
Catalog No. 50-210-6197 Supplier ACROBiosystems Supplier No. EGRHP2E3200TESTS
Recombinant Protein;200TESTS ;PE-Labeled Human EGF R, His Tag (EGR-HP2E3) is produced via site-specific conjugation of PE to Human EGF R (25-645), His Tag under optimal conditions with a proprietary technology. Human EGF R (25-645), His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.3 kDa.